siRNA Details
Gene Symbol: D930020B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177335.4 | NP_796309.3 | 13 | 2494 | ||
GenBank mRNA | AK045823.1 | - | 1 | 204 | ||
AK086308.1 | - | 1 | 562 | |||
BB644359.1 | - | 1 | 202 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
