siRNA Details
Gene Symbol: Dnah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033668.1 | NP_001028840.1 | 67 | 10698 | ||
GenBank mRNA | AK046346.1 | - | 9 | 1323 | ||
AK052631.1 | - | 9 | 1325 | |||
AK163552.1 | - | 9 | 1323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
