siRNA Details
Gene Symbol: Trpm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035239.2 | NP_001030316.1 | 11 | 1548 | ||
NM_001035240.2 | NP_001030317.1 | 11 | 1548 | |||
NM_001035241.2 | NP_001030318.1 | 11 | 1548 | |||
NM_001035242.1 | NP_001030319.1 | 11 | 1548 | |||
NM_001035243.2 | NP_001030320.1 | 11 | 1548 | |||
NM_177341.4 | NP_796315.2 | 11 | 1615 | |||
GenBank mRNA | AJ544532.1 | - | 11 | 1548 | ||
AJ544533.1 | - | 11 | 1548 | |||
AJ544534.1 | - | 11 | 1548 | |||
AJ544535.1 | - | 11 | 1548 | |||
AJ544536.1 | - | 11 | 1548 | |||
AK032465.1 | - | 1 | 1012 | |||
AK044454.1 | - | 11 | 1617 | |||
AK051867.1 | - | 6 | 681 | |||
AK142504.1 | - | 12 | 1504 | |||
AK173218.1 | - | 11 | 1619 | |||
BC141400.1 | - | 11 | 1591 | 9056013 | ||
BC141412.1 | - | 11 | 1591 | 9056025 | ||
HM064407.1 | - | 11 | 1640 | |||
HM064408.1 | - | 11 | 1640 | |||
HM064409.1 | - | 11 | 1640 | |||
HM064410.1 | - | 12 | 1715 | |||
JF706722.1 | - | 11 | 1611 | |||
JX644970.1 | - | 11 | 1640 | |||
JX644971.1 | - | 11 | 1640 | |||
JX644972.1 | - | 11 | 1640 | |||
JX644973.1 | - | 11 | 1640 | |||
JX644974.1 | - | 11 | 1640 | |||
JX644975.1 | - | 12 | 1715 | |||
JX644979.1 | - | 12 | 1715 | |||
JX644980.1 | - | 12 | 1715 | |||
JX644981.1 | - | 12 | 1715 | |||
JX644983.1 | - | 11 | 1640 | |||
JX644984.1 | - | 11 | 1640 | |||
JX644985.1 | - | 11 | 1640 | |||
JX644986.1 | - | 11 | 1640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
