siRNA Details
Gene Symbol: Zbtb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177660.3 | NP_808328.2 | 6 | 3013 | ||
GenBank mRNA | AK048280.1 | - | 5 | 1475 | ||
AK083443.1 | - | 4 | 624 |

More Information
Other Species Targeted

Gene Ontology Categories:
