siRNA Details
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177680.3 | NP_808348.2 | 10 | 1460 | ||
GenBank mRNA | AB221619.1 | - | 2 | 84 | ||
AI121974.1 | - | 3 | 195 | |||
AK040341.1 | - | 1 | 3577 | |||
AK173312.1 | - | 7 | 723 | |||
BC022697.1 | - | 3 | 253 | |||
BC037593.1 | - | 4 | 353 | |||
BC044650.1 | - | 8 | 1278 | |||
BC048817.1 | - | 3 | 238 | |||
CO424360.1 | - | 5 | 405 |

More Information
Other Species Targeted

Gene Ontology Categories:
