siRNA Details
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177680.3 | NP_808348.2 | 10 | 1492 | ||
GenBank mRNA | AB221619.1 | - | 2 | 116 | ||
AI121974.1 | - | 3 | 227 | |||
AK040341.1 | - | 1 | 3609 | |||
AK173312.1 | - | 7 | 755 | |||
BC022697.1 | - | 3 | 285 | |||
BC037593.1 | - | 4 | 385 | |||
BC044650.1 | - | 8 | 1310 | |||
BC048817.1 | - | 3 | 270 | |||
CO424360.1 | - | 5 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
