siRNA Details
Gene Symbol: Ythdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177680.3 | NP_808348.2 | 11 | 1591 | ||
GenBank mRNA | AB221619.1 | - | 3 | 215 | ||
AI121974.1 | - | 4 | 326 | |||
AK040341.1 | - | 2 | 3708 | |||
AK173312.1 | - | 8 | 854 | |||
BC022697.1 | - | 4 | 384 | |||
BC037593.1 | - | 5 | 484 | |||
BC044650.1 | - | 9 | 1409 | |||
BC048817.1 | - | 4 | 369 | |||
CO424360.1 | - | 6 | 536 |

More Information
Other Species Targeted

Gene Ontology Categories:
