siRNA Details
Gene Symbol: Mapk1ip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178684.5 | NP_848799.1 | 4 | 1087 | ||
GenBank mRNA | AK033615.1 | - | 2 | 644 | ||
AK048061.1 | - | 4 | 1125 | |||
AK086896.1 | - | 4 | 1115 | |||
AK143495.1 | - | 3 | 1012 | |||
AK168694.1 | - | 4 | 1118 | |||
BC089366.1 | - | 2 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
