siRNA Details
Gene Symbol: Rims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK220335.1 | - | 4745 | |||
BC016274.1 | - | 1558 | ||||
BC024828.1 | - | 1405 | ||||
BC037619.1 | - | 1576 | ||||
BC055294.1 | - | 1525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
