siRNA Details
Gene Symbol: Rims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK220335.1 | - | 5505 | |||
BC037619.1 | - | 2336 | ||||
BC055294.1 | - | 2291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
