siRNA Details
Gene Symbol: Nrde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290303.1 | NP_001277232.1 | 11 | 2434 | ||
NM_183155.3 | NP_898978.2 | 11 | 2419 | |||
GenBank mRNA | BC002230.1 | - | 1 | 128 | ||
BC025577.1 | - | 6 | 1039 | 5323598 | ||
BC051175.1 | - | 10 | 2324 | |||
DN174402.1 | - | 2 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
