siRNA Details
Gene Symbol: Nrde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290303.1 | NP_001277232.1 | 12 | 3302 | ||
NM_183155.3 | NP_898978.2 | 12 | 3287 | |||
GenBank mRNA | BC025577.1 | - | 7 | 1907 | 5323598 | |
BC051175.1 | - | 11 | 3192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
