siRNA Details
Gene Symbol: Mfsd7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081259.1 | NP_001074728.1 | 1 | 428 | ||
GenBank mRNA | AK036315.1 | - | 1 | 206 | ||
AK165872.1 | - | 1 | 427 | |||
BC010797.1 | - | 1 | 362 | |||
BC157992.1 | - | 1 | 396 | 9007305 |

More Information
Other Species Targeted

Gene Ontology Categories:
