siRNA Details
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 18 | 1973 | ||
NM_001291444.1 | NP_001278373.1 | 18 | 1979 | |||
NM_001291445.1 | NP_001278374.1 | 17 | 1883 | |||
GenBank mRNA | AK030003.1 | - | 13 | 1342 | ||
AK036581.1 | - | 17 | 1859 | |||
AK054000.1 | - | 18 | 1954 | |||
AK122326.1 | - | 16 | 1662 | |||
BC059076.1 | - | 3 | 543 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
