siRNA Details
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 24 | 2806 | ||
NM_001291444.1 | NP_001278373.1 | 24 | 2812 | |||
NM_001291445.1 | NP_001278374.1 | 23 | 2716 | |||
GenBank mRNA | AK030003.1 | - | 19 | 2175 | ||
AK036581.1 | - | 23 | 2692 | |||
AK054000.1 | - | 24 | 2787 | |||
AK122326.1 | - | 22 | 2495 | |||
BC043302.1 | - | 5 | 723 | |||
BC059076.1 | - | 9 | 1376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
