siRNA Details
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 24 | 5353 | ||
NM_001291444.1 | NP_001278373.1 | 24 | 5359 | |||
NM_001291445.1 | NP_001278374.1 | 23 | 5263 | |||
GenBank mRNA | AK122326.1 | - | 22 | 5044 | ||
AK216671.1 | - | 1 | 217 | |||
BC026674.1 | - | 1 | 893 | |||
BC043302.1 | - | 6 | 3184 | |||
BC051499.1 | - | 1 | 659 | |||
BC059076.1 | - | 10 | 3835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
