siRNA Details
Gene Symbol: Phactr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005740.1 | NP_001005740.1 | 7 | 1278 | ||
GenBank mRNA | AK048208.1 | - | 7 | 1174 | ||
AK139452.1 | - | 7 | 1250 | |||
AY993932.1 | - | 8 | 1215 | |||
BC043065.1 | - | 2 | 304 | |||
BC048407.1 | - | 6 | 1109 | |||
BC061691.1 | - | 7 | 1278 | 6307669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
