siRNA Details
Gene Symbol: Samd9l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010156.3 | NP_034286.2 | 2 | 3249 | ||
GenBank mRNA | AK032210.1 | - | 1 | 827 | ||
AK035419.1 | - | 1 | 1120 | |||
AK052535.1 | - | 2 | 3251 | |||
AK172649.1 | - | 2 | 3136 | |||
AK173329.1 | - | 2 | 3117 | |||
BC031151.1 | - | 1 | 1082 | |||
BC079894.1 | - | 2 | 3170 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
