siRNA Details
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019587.2 | NP_062533.2 | 2 | 1016 | ||
GenBank mRNA | AK138631.1 | - | 2 | 91 | ||
AK140240.1 | - | 2 | 1020 |

More Information
Other Species Targeted

Gene Ontology Categories:
