siRNA Details
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019587.2 | NP_062533.2 | 13, 14 | 2535 | ||
GenBank mRNA | AK122467.1 | - | 12, 13 | 1461 | ||
AK138168.1 | - | 11, 12 | 1362 | |||
AK140240.1 | - | 13, 14 | 2539 |

More Information
Other Species Targeted

Gene Ontology Categories:
