siRNA Details
Gene Symbol: Rnf187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022423.2 | NP_071868.2 | 4 | 1739 | ||
GenBank mRNA | AB030190.1 | - | 4 | 1731 | ||
AK037991.1 | - | 4 | 1654 | |||
AK075944.1 | - | 4 | 1751 | |||
AK076158.1 | - | 4 | 1729 | |||
AK088973.1 | - | 4 | 1742 | |||
BC072613.1 | - | 4 | 1732 | |||
BC096522.1 | - | 4 | 1703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
