siRNA Details
Gene Symbol: Pgd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081274.1 | NP_001074743.1 | 13 | 2077 | ||
GenBank mRNA | AK002894.1 | - | 13 | 2039 | ||
AK004217.1 | - | 6 | 1270 | |||
AK077110.1 | - | 13 | 2036 | |||
AK145602.1 | - | 13 | 2041 | |||
AK150210.1 | - | 13 | 2040 | |||
AK150490.1 | - | 13 | 2036 | |||
AK151380.1 | - | 13 | 2036 | |||
AK151993.1 | - | 13 | 2036 | |||
AK152758.1 | - | 13 | 2036 | |||
AK152894.1 | - | 13 | 2036 | |||
AK153409.1 | - | 13 | 2033 | |||
AK155027.1 | - | 13 | 2036 | |||
AK166733.1 | - | 13 | 2031 | |||
AK166947.1 | - | 13 | 2013 | |||
AK167215.1 | - | 13 | 2031 | |||
AK168251.1 | - | 13 | 2036 | |||
AK198787.1 | - | 1 | 191 | |||
AK203964.1 | - | 1 | 44 | |||
AK205810.1 | - | 1 | 191 | |||
AK208960.1 | - | 1 | 202 | |||
BC008646.1 | - | 13 | 2008 | 3155147 | ||
BC011329.1 | - | 13 | 2026 | 3984654 | ||
BC014793.1 | - | 13 | 1845 | 3155100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
