siRNA Details
Gene Symbol: Ms4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025610.4 | NP_001020781.1 | 6 | 921 | ||
NM_001276398.1 | NP_001263327.1 | 7 | 1218 | |||
NM_027836.6 | NP_082112.3 | 7 | 978 | |||
GenBank mRNA | AK079888.1 | - | 7 | 1219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
