siRNA Details
Gene Symbol: Pwp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029546.2 | NP_083822.1 | 5 | 453 | ||
GenBank mRNA | AB041855.1 | - | 5 | 414 | ||
AK088254.1 | - | 5 | 454 | |||
AK088983.1 | - | 5 | 454 | |||
AK196838.1 | - | 1 | 19 | |||
BC112325.1 | - | 5 | 419 | 5355810 | ||
BC117903.1 | - | 5 | 434 | |||
BC128480.1 | - | 5 | 434 | 40097069 |
