siRNA Details
Gene Symbol: Npnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287101.1 | NP_001274030.1 | 5 | 644 | ||
NM_001287102.1 | NP_001274031.1 | 4 | 593 | |||
GenBank mRNA | AF397007.1 | - | 4 | 345 | ||
AF397008.1 | - | 5 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
