siRNA Details
Gene Symbol: TUBA3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006001.2 | NP_005992.1 | 3 | 401 | ||
GenBank mRNA | BC011721.2 | - | 3 | 344 | 4025942 | |
DB026664.1 | - | 3 | 401 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
