siRNA Details
Gene Symbol: Dok4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053246.3 | NP_444476.1 | 9 | 2318 | ||
GenBank mRNA | AK077612.1 | - | 9 | 2311 | ||
AK139363.1 | - | 8 | 2209 | |||
AK142621.1 | - | 8 | 2225 | |||
AK156485.1 | - | 9 | 2318 | |||
BC004705.1 | - | 9 | 2279 | 3499419 |

More Information
Other Species Targeted

Gene Ontology Categories:
