siRNA Details
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054055.2 | NP_473396.1 | 13 | 2273 | ||
GenBank mRNA | AF306491.1 | - | 13 | 2272 | ||
AK085326.1 | - | 13 | 2274 | |||
AK087518.1 | - | 13 | 2275 | |||
AK128982.1 | - | 13 | 2270 | |||
AK138344.1 | - | 13 | 2271 | |||
AK157881.1 | - | 13 | 2244 | |||
AK165355.1 | - | 13 | 2277 | |||
BC026803.1 | - | 13 | 2259 | 4238769 | ||
BC028882.1 | - | 12 | 2134 |
