siRNA Details
Gene Symbol: Synpo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080451.2 | NP_536699.2 | 4 | 1343 | ||
GenBank mRNA | AJ306625.1 | - | 1 | 211 | ||
BC158045.1 | - | 4 | 1290 | 9007349 |

More Information
Other Species Targeted

Gene Ontology Categories:
