siRNA Details
Gene Symbol: Cntnap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130457.2 | NP_569724.2 | 7 | 1111 | ||
GenBank mRNA | AF333770.1 | - | 7 | 1076 | ||
AK042607.1 | - | 7 | 1095 | |||
AK053611.1 | - | 7 | 1103 | |||
AK081891.1 | - | 7 | 1104 | |||
AK142816.1 | - | 7 | 1107 | |||
AK173255.1 | - | 7 | 1086 | |||
BC034628.1 | - | 7 | 1111 | 5369648 |
