siRNA Details
Gene Symbol: Baiap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK170190.1 | - | 7 | 688 | ||
BC006620.1 | - | 13 | 1637 | 3158004 | ||
BC016411.1 | - | 13 | 1637 | 3158004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
