siRNA Details
Gene Symbol: Baiap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037754.3 | NP_001032843.1 | 14 | 1857 | ||
NM_130862.4 | NP_570932.2 | 14 | 1977 | |||
GenBank mRNA | AK083201.1 | - | 1 | 72 | ||
AK145924.1 | - | 14 | 1978 | |||
AK158579.1 | - | 14 | 1999 | |||
AK160401.1 | - | 14 | 1859 | |||
AK170190.1 | - | 7 | 1506 | |||
BC006620.1 | - | 13 | 2455 | 3158004 | ||
BC016411.1 | - | 13 | 2455 | 3158004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
