siRNA Details
Gene Symbol: Apcdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB023957.1 | - | 244 | |||
AK157407.1 | - | 1505 | ||||
AK172004.1 | - | 2942 | ||||
BC065780.1 | - | 217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
