siRNA Details
Gene Symbol: Fam208b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134063.3 | NP_598824.3 | 14 | 5541 | ||
GenBank mRNA | AK128934.1 | - | 1 | 858 | ||
AK131707.1 | - | 3 | 2937 | |||
AK165123.1 | - | 1 | 1724 | |||
AK220437.1 | - | 14 | 5529 | |||
BC016423.1 | - | 1 | 225 | |||
BC045617.1 | - | 1 | 564 | |||
BC085160.1 | - | 5 | 4284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
