siRNA Details
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037955.4 | NP_001033044.1 | 7 | 2815 | ||
NM_134068.3 | NP_598829.1 | 8 | 859 | |||
GenBank mRNA | AF237619.1 | - | 8 | 819 | ||
AK003947.1 | - | 1 | 448 | |||
BY477408.1 | - | 1 | 266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
