siRNA Details
Gene Symbol: Rcbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170694.1 | NP_001164165.1 | 11 | 1273 | ||
NM_134083.4 | NP_598844.3 | 11 | 1268 | |||
GenBank mRNA | AK013540.1 | - | 1378 | |||
AK031242.1 | - | 11 | 1234 | |||
AK031248.1 | - | 11 | 1239 | |||
AK147784.1 | - | 12 | 1274 | |||
AK160881.1 | - | 12 | 1460 | |||
BC003224.1 | - | 9 | 1066 | 3482729 | ||
JN086338.1 | - | 10 | 1189 | |||
JN086339.1 | - | 11 | 1427 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
