siRNA Details
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134123.3 | NP_598884.2 | 7 | 1037 | ||
GenBank mRNA | AK054357.1 | - | 1 | 967 | ||
AK122451.1 | - | 7 | 904 | |||
AK157957.1 | - | 7 | 1043 | |||
AK171078.1 | - | 8 | 844 | |||
AK200783.1 | - | 3 | 217 | |||
AK218201.1 | - | 3 | 217 | |||
BC038363.1 | - | 7 | 681 | 5371946 | ||
BC075621.1 | - | 7 | 1030 | 6406441 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
