siRNA Details
Gene Symbol: Carns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134148.2 | NP_598909.2 | 10 | 2564 | ||
GenBank mRNA | AK129347.1 | - | 9 | 2721 | ||
BC019736.1 | - | 1 | 286 | |||
BC023699.1 | - | 2 | 978 | 5345551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
