siRNA Details
Gene Symbol: Fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286424.1 | NP_001273353.1 | 4 | 2291 | ||
NM_134161.3 | NP_598922.1 | 4 | 2398 | |||
GenBank mRNA | BC030474.1 | - | 4 | 2295 | 5368333 | |
BC062113.1 | - | 4 | 2397 | 6816067 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
