siRNA Details
Gene Symbol: Vmn1r237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134200.1 | NP_598961.1 | 1 | 489 | ||
GenBank mRNA | BC125543.1 | - | 1 | 543 | 40130058 | |
BC138066.1 | - | 1 | 543 | 8861085 |
