siRNA Details
Gene Symbol: Bcas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166642.1 | NP_001160114.1 | 7 | 511 | ||
NM_001166643.1 | NP_001160115.1 | 6 | 425 | |||
NM_138681.4 | NP_619622.3 | 7 | 511 | |||
GenBank mRNA | AF313800.1 | - | 6 | 405 | ||
AK032423.1 | - | 7 | 462 | |||
AK034117.1 | - | 7 | 491 | |||
AK157222.1 | - | 7 | 462 | |||
AK160127.1 | - | 6 | 404 | |||
AK171964.1 | - | 6 | 378 | |||
BC028339.1 | - | 6 | 399 | |||
BC038173.1 | - | 5 | 368 | |||
BC050233.1 | - | 6 | 404 | |||
BC060690.1 | - | 6 | 388 | |||
BC080775.1 | - | 5 | 372 | |||
BC086693.1 | - | 6 | 392 | |||
BC118965.2 | - | 6 | 425 | 40097104 | ||
BC118966.1 | - | 6 | 407 | 40097105 |
