siRNA Details
Gene Symbol: Bcas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166642.1 | NP_001160114.1 | 14 | 1225 | ||
NM_001166643.1 | NP_001160115.1 | 13 | 1139 | |||
NM_138681.4 | NP_619622.3 | 14 | 1225 | |||
GenBank mRNA | AK032423.1 | - | 14 | 1176 | ||
AK034117.1 | - | 14 | 1205 | |||
AK157222.1 | - | 14 | 1175 | |||
AK160127.1 | - | 13 | 1118 | |||
AK171964.1 | - | 13 | 1092 | |||
BC028339.1 | - | 13 | 1113 | |||
BC038173.1 | - | 12 | 1082 | |||
BC050233.1 | - | 13 | 1118 | |||
BC060690.1 | - | 13 | 1102 | |||
BC080775.1 | - | 12 | 1086 | |||
BC086693.1 | - | 13 | 1106 | |||
BC118965.2 | - | 13 | 1139 | 40097104 | ||
BC118966.1 | - | 13 | 1121 | 40097105 |
