siRNA Details
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138953.2 | NP_620403.2 | 11, 12 | 1975 | ||
GenBank mRNA | AK140906.1 | - | 11, 12 | 1929 | ||
AK144197.1 | - | 11, 12 | 1940 | |||
AK144978.1 | - | 11, 12 | 1939 | |||
AK145868.1 | - | 11, 12 | 1924 | |||
AK154545.1 | - | 11, 12 | 1936 | |||
AK156164.1 | - | 11, 12 | 1942 | |||
AK161961.1 | - | 4, 5 | 425 | |||
AK163470.1 | - | 1, 2 | 2801 | |||
BC006925.1 | - | 6, 7 | 1007 | |||
BC151188.1 | - | 11, 12 | 1930 | 9088077 |

More Information
Other Species Targeted

Gene Ontology Categories:
