siRNA Details
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138953.2 | NP_620403.2 | 12 | 2078 | ||
GenBank mRNA | AK144197.1 | - | 12 | 2043 | ||
AK144978.1 | - | 12 | 2042 | |||
AK145868.1 | - | 12 | 2027 | |||
AK156164.1 | - | 12 | 2045 | |||
AK161961.1 | - | 5 | 528 | |||
BC006925.1 | - | 7 | 1110 | |||
BC151188.1 | - | 12 | 2033 | 9088077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
