siRNA Details
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138953.2 | NP_620403.2 | 12 | 2832 | ||
GenBank mRNA | AK144197.1 | - | 12 | 2843 | ||
AK145868.1 | - | 12 | 2781 | |||
AK156164.1 | - | 12 | 2834 | |||
AK161961.1 | - | 5 | 1286 | |||
BC006925.1 | - | 7 | 1910 | |||
BC151188.1 | - | 12 | 2833 | 9088077 |

More Information
Other Species Targeted

Gene Ontology Categories:
