siRNA Details
Gene Symbol: Metrnl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144797.3 | NP_659046.1 | 4 | 1150 | ||
GenBank mRNA | AK034855.1 | - | 4 | 1151 | ||
AK085880.1 | - | 4 | 1152 | |||
AK170549.1 | - | 3 | 765 | |||
AK170800.1 | - | 4 | 1152 | |||
AK171200.1 | - | 4 | 896 | |||
BC019776.1 | - | 4 | 1105 | 3672224 | ||
BC024445.1 | - | 4 | 1165 | 4974030 | ||
BC024497.1 | - | 1153 | 4982563 | |||
BC026646.1 | - | 4 | 1144 | 4987595 |
