siRNA Details
Gene Symbol: Plekha5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC010246.1 | - | 1 | 2824 | ||
BC014853.1 | - | 17 | 2922 | 3598156 | ||
BC021505.1 | - | 16 | 2609 | 5347087 | ||
BC025596.1 | - | 10 | 1639 | |||
BC031390.1 | - | 16 | 2729 | |||
BC033481.1 | - | 19 | 3439 | 4945488 |
