siRNA Details
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 17, 18 | 1842 | ||
NM_001285894.1 | NP_001272823.1 | 18, 19 | 1795 | |||
GenBank mRNA | AK029007.1 | - | 1, 2 | 58 | ||
AK122375.1 | - | 18, 19 | 1744 | |||
AK141678.1 | - | 17, 18 | 1765 | |||
BC023914.1 | - | 3, 4 | 233 | 5323126 | ||
BC055323.1 | - | 17, 18 | 1759 | 5706288 |

More Information
Other Species Targeted

Gene Ontology Categories:
