siRNA Details
Gene Symbol: Vps8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285893.1 | NP_001272822.1 | 22 | 2171 | ||
NM_001285894.1 | NP_001272823.1 | 23 | 2124 | |||
GenBank mRNA | AK029007.1 | - | 6 | 387 | ||
AK122375.1 | - | 23 | 2073 | |||
AK141678.1 | - | 22 | 2094 | |||
BC023914.1 | - | 8 | 562 | 5323126 | ||
BC032214.1 | - | 2 | 140 | 5323693 | ||
BC055323.1 | - | 22 | 2088 | 5706288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
