siRNA Details
Gene Symbol: Phyhip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145981.3 | NP_666093.1 | 5 | 2086 | ||
GenBank mRNA | AK049900.1 | - | 5 | 2087 | ||
BC030494.1 | - | 5 | 2083 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
